COM448668051 Combi-Blocks, Inc.

Lithium trifluoro(trifluoromethyl)borate-dimethyl carbonate complex

  • MDL number MFCD22124722
  • Cas Number : 1443685-69-5
  • Molecular Weight : 233.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0035
Availability details for
Quantity
  • Availability
  • Add to Cart