COM448668056 Combi-Blocks, Inc.

2-(4-Hydroxyphenyl)-4,5-diphenylimidazole

  • MDL number MFCD00740542
  • Cas Number : 1752-94-9
  • Molecular Weight : 312.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0138
Availability details for
Quantity
  • Availability
  • Add to Cart