COM448668064 Combi-Blocks, Inc.

2,6-Bis[(2,2,6,6-tetramethyl-1-piperidinyl)methyl]phenylboronic acid

  • MDL number MFCD22581436
  • Cas Number : 1243264-54-1
  • Molecular Weight : 428.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0319
Availability details for
Quantity
  • Availability
  • Add to Cart