COM448668078 Combi-Blocks, Inc.

4-(Cis-4-hydroxycyclohexyl)phenol

  • MDL number MFCD23380209
  • Cas Number : 370860-74-5
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0668
Availability details for
Quantity
  • Availability
  • Add to Cart