COM448668080 Combi-Blocks, Inc.

Diethyl 3,3'-iminodipropionate

  • MDL number MFCD14705114
  • Cas Number : 3518-88-5
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0721
Availability details for
Quantity
  • Availability
  • Add to Cart