COM448668087 Combi-Blocks, Inc.

4-Bromo-n,n,3,5-tetramethylaniline

  • MDL number MFCD11036245
  • Cas Number : 14275-09-3
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0965
Availability details for
Quantity
  • Availability
  • Add to Cart