COM448668088 Combi-Blocks, Inc.

4-Methoxyphenyl 4'-(3-butenyloxy)benzoate

  • MDL number MFCD00143337
  • Cas Number : 76487-56-4
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0973
Availability details for
Quantity
  • Availability
  • Add to Cart