COM448668089 Combi-Blocks, Inc.

Isorhapontigenin

  • MDL number MFCD12407151
  • Cas Number : 32507-66-7
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1012
Availability details for
Quantity
  • Availability
  • Add to Cart