COM448668090 Combi-Blocks, Inc.

9,9-Bis(4-bromophenyl)fluorene

  • MDL number MFCD19707068
  • Cas Number : 128406-10-0
  • Molecular Weight : 476.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1036
Availability details for
Quantity
  • Availability
  • Add to Cart