COM448668102 Combi-Blocks, Inc.

Methyl 4,6-o-benzylidene-2-deoxy-alpha-d-erythro-hexopyranosid-3-ulose oxime

  • MDL number MFCD15072181
  • Cas Number : 63598-32-3
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1260
Availability details for
Quantity
  • Availability
  • Add to Cart