COM448668110 Combi-Blocks, Inc.

Trimethyl[2-[(trimethylsilyl)methyl]benzyl]ammonium iodide

  • MDL number MFCD12407129
  • Cas Number : 83781-47-9
  • Molecular Weight : 363.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1510
Availability details for
Quantity
  • Availability
  • Add to Cart