COM448668112 Combi-Blocks, Inc.

2,6-Bis(hydroxymethyl)naphthalene

  • MDL number MFCD11870109
  • Cas Number : 5859-93-8
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1566
Availability details for
Quantity
  • Availability
  • Add to Cart