COM448668118 Combi-Blocks, Inc.

(R)-4-(4-Methoxybenzyloxy)-1,2-butanediol

  • MDL number MFCD11110643
  • Cas Number : 213978-61-1
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1610
Availability details for
Quantity
  • Availability
  • Add to Cart