COM448668121 Combi-Blocks, Inc.

4-Iodo-4',4''-dimethoxytriphenylamine

  • MDL number MFCD23702008
  • Cas Number : 201802-15-5
  • Molecular Weight : 431.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1758
Availability details for
Quantity
  • Availability
  • Add to Cart