COM448668123 Combi-Blocks, Inc.

O-Acetyl-n-carbobenzoxyhydroxylamine

  • MDL number MFCD11975425
  • Cas Number : 180798-01-0
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-1771
Availability details for
Quantity
  • Availability
  • Add to Cart