COM448668133 Combi-Blocks, Inc.

4-Bromo-1,2-bis-dibromomethyl-benzene

  • MDL number MFCD00465850
  • Cas Number : 4235-46-5
  • Molecular Weight : 500.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2015
Availability details for
Quantity
  • Availability
  • Add to Cart