COM448668139 Combi-Blocks, Inc.

1-Bromo-3-phenylimidazo[1,5-a]pyridine

  • MDL number MFCD04587511
  • Cas Number : 104202-15-5
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2103
Availability details for
Quantity
  • Availability
  • Add to Cart