COM448668140 Combi-Blocks, Inc.

2-(3-Thienyl)-1,3-dioxolane

  • MDL number MFCD06209359
  • Cas Number : 13250-82-3
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2135
Availability details for
Quantity
  • Availability
  • Add to Cart