COM448668145 Combi-Blocks, Inc.

Gal[26bn]beta(1-4)glc[236bn]-beta-mp

  • MDL number MFCD11112170
  • Cas Number : 358681-61-5
  • Molecular Weight : 899.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2261
Availability details for
Quantity
  • Availability
  • Add to Cart