COM448668147 Combi-Blocks, Inc.

2,4,6-Tri(9h-carbazol-9-yl)-1,3,5-triazine

  • MDL number MFCD18207722
  • Cas Number : 134984-37-5
  • Molecular Weight : 576.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2309
Availability details for
Quantity
  • Availability
  • Add to Cart