COM448668151 Combi-Blocks, Inc.

(S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane

  • MDL number MFCD22041647
  • Cas Number : 151223-12-0
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2438
Availability details for
Quantity
  • Availability
  • Add to Cart