COM448668154 Combi-Blocks, Inc.

3,4-Difluoro-2,5-bis(trimethylsilyl)thiophene

  • MDL number MFCD11036246
  • Cas Number : 347838-12-4
  • Molecular Weight : 264.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2514
Availability details for
Quantity
  • Availability
  • Add to Cart