COM448668162 Combi-Blocks, Inc.

Pentadecanophenone

  • MDL number MFCD20257914
  • Cas Number : 4669-04-9
  • Molecular Weight : 302.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2766
Availability details for
Quantity
  • Availability
  • Add to Cart