COM448668176 Combi-Blocks, Inc.

Phenyl 3-o-allyl-2,4,6-tri-o-benzyl-1-thio-beta-d-galactopyranoside

  • MDL number MFCD11112183
  • Cas Number : 1017587-57-3
  • Molecular Weight : 582.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3072
Availability details for
Quantity
  • Availability
  • Add to Cart