COM448668177 Combi-Blocks, Inc.

Decahydro-2,6-naphthalenedicarboxylic acid dimethyl ester

  • MDL number MFCD11042680
  • Cas Number : 3068-02-8
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3080
Availability details for
Quantity
  • Availability
  • Add to Cart