COM448668180 Combi-Blocks, Inc.

1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic acid sesquihydrate

  • MDL number MFCD15478075
  • Cas Number : 84294-96-2
  • Molecular Weight : 658.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3152
Availability details for
Quantity
  • Availability
  • Add to Cart