COM448668192 Combi-Blocks, Inc.

1,3-Dibromo-7-tert-butylpyrene

  • MDL number MFCD22581440
  • Cas Number : 1005771-04-9
  • Molecular Weight : 416.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3541
Availability details for
Quantity
  • Availability
  • Add to Cart