COM448668196 Combi-Blocks, Inc.

Dimethyl trans-1,2-cyclohexanedicarboxylate

  • MDL number MFCD09998658
  • Cas Number : 3205-35-4
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3635
Availability details for
Quantity
  • Availability
  • Add to Cart