COM448668202 Combi-Blocks, Inc.

3-Methoxyflavone

  • MDL number MFCD00017612
  • Cas Number : 7245-02-5
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3782

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3782
Availability details for
Quantity
  • Availability
  • Add to Cart