COM448668203 Combi-Blocks, Inc.

Di-mu-chlorobis[2-[(dimethylamino)methyl]-4,6-dimethoxyphenyl-c,n]dipalladium(ii)

  • MDL number MFCD22124719
  • Cas Number : 18987-71-8
  • Molecular Weight : 672.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3790
Availability details for
Quantity
  • Availability
  • Add to Cart