COM448668205 Combi-Blocks, Inc.

6,7-Dihydroxycoumarin-3-carboxylic acid

  • MDL number MFCD18450959
  • Cas Number : 84738-35-2
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3808
Availability details for
Quantity
  • Availability
  • Add to Cart