COM448668209 Combi-Blocks, Inc.

2-Butyl-6-phenylpyridine

  • MDL number MFCD00796985
  • Cas Number : 60272-71-1
  • Molecular Weight : 211.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3954
Availability details for
Quantity
  • Availability
  • Add to Cart