COM448668211 Combi-Blocks, Inc.

N-(5-Aminopentyl)acetamide

  • MDL number MFCD19204554
  • Cas Number : 32343-73-0
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3990

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-3990
Availability details for
Quantity
  • Availability
  • Add to Cart