COM448668215 Combi-Blocks, Inc.

1,3-Di-tert-butyl-1,3,2-diazaphospholidine 2-oxide

  • MDL number MFCD14708171
  • Cas Number : 854929-38-7
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4103
Availability details for
Quantity
  • Availability
  • Add to Cart