COM448668219 Combi-Blocks, Inc.

8-Azahypoxanthine

  • MDL number MFCD00005804
  • Cas Number : 2683-90-1
  • Molecular Weight : 137.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4159
Availability details for
Quantity
  • Availability
  • Add to Cart