COM448668220 Combi-Blocks, Inc.

Dimethyl 2-(1,3-dithiole)phosphonate

  • MDL number MFCD20265584
  • Cas Number : 133113-76-5
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4161

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4161
Availability details for
Quantity
  • Availability
  • Add to Cart