COM448668222 Combi-Blocks, Inc.

7-Methoxycoumarin-3-carboxylic acid n-succinimidyl ester

  • MDL number MFCD00467593
  • Cas Number : 150321-92-9
  • Molecular Weight : 317.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4200
Availability details for
Quantity
  • Availability
  • Add to Cart