COM448668224 Combi-Blocks, Inc.

Trans-2,3-dichloro-1,4-dioxane

  • MDL number MFCD00191421
  • Cas Number : 3883-43-0
  • Molecular Weight : 157.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4252
Availability details for
Quantity
  • Availability
  • Add to Cart