COM448668227 Combi-Blocks, Inc.

4,4'-Bis(chloromethyl)-2,2'-bipyridyl

  • MDL number MFCD20265579
  • Cas Number : 138219-98-4
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4326
Availability details for
Quantity
  • Availability
  • Add to Cart