COM448668228 Combi-Blocks, Inc.

Sodium 4-aminophenylarsonate

  • MDL number MFCD00067431
  • Cas Number : 127-85-5
  • Molecular Weight : 261.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4344
Availability details for
Quantity
  • Availability
  • Add to Cart