COM448668229 Combi-Blocks, Inc.

1,3-Dihydro-1-methoxyisobenzofuran

  • MDL number MFCD12407127
  • Cas Number : 67536-29-2
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4348
Availability details for
Quantity
  • Availability
  • Add to Cart