COM448668230 Combi-Blocks, Inc.

4-Methoxyphenyl 4-o-acetyl-3,6-di-o-benzyl-2-deoxy-2-phthalimido-beta-d-glucopyranoside

  • MDL number MFCD11112179
  • Cas Number : 140615-77-6
  • Molecular Weight : 637.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4382
Availability details for
Quantity
  • Availability
  • Add to Cart