COM448668234 Combi-Blocks, Inc.

Trans-4-propylcyclohexanol

  • MDL number MFCD11114411
  • Cas Number : 77866-58-1
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4424
Availability details for
Quantity
  • Availability
  • Add to Cart