COM448668242 Combi-Blocks, Inc.

(S)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5h-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate

  • MDL number MFCD23380202
  • Molecular Weight : 431.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4597
Availability details for
Quantity
  • Availability
  • Add to Cart