COM448668248 Combi-Blocks, Inc.

3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide

  • MDL number MFCD04038410
  • Cas Number : 176650-93-4
  • Molecular Weight : 503.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4693
Availability details for
Quantity
  • Availability
  • Add to Cart