COM448668254 Combi-Blocks, Inc.

5-Amino-n,n'-bis(2,3-dihydroxypropyl)isophthalamide HCl

  • MDL number MFCD00137747
  • Cas Number : 203515-86-0
  • Molecular Weight : 363.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4837
Availability details for
Quantity
  • Availability
  • Add to Cart