COM448668257 Combi-Blocks, Inc.

(4-Ethynyl-phenyl)-diphenyl-amine

  • MDL number MFCD09030769
  • Cas Number : 205877-26-5
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4894
Availability details for
Quantity
  • Availability
  • Add to Cart