COM448668263 Combi-Blocks, Inc.

(1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid

  • MDL number MFCD20259654
  • Cas Number : 1259278-10-8
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4977

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4977
Availability details for
Quantity
  • Availability
  • Add to Cart