COM448668265 Combi-Blocks, Inc.

Hexahydroxybenzene

  • MDL number MFCD00013966
  • Cas Number : 608-80-0
  • Molecular Weight : 174.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4990

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-4990
Availability details for
Quantity
  • Availability
  • Add to Cart