COM448668273 Combi-Blocks, Inc.

5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene

  • MDL number MFCD23380207
  • Cas Number : 170702-05-3
  • Molecular Weight : 492.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5132
Availability details for
Quantity
  • Availability
  • Add to Cart